Anti-Interleukin 17 Rabbit Polyclonal Antibody
USBII8439-04P
- Antibody type:Primary
- Antigen name:Interleukin 17
- Antigen symbol:IL17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH. 7.2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Interleukin 17
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse