Order Entry
Export
ContactUsLinkComponent
Anti-Interleukin 15 Receptor alpha Goat Polyclonal Antibody (Carboxyfluorescein)
Anti-Interleukin 15 Receptor alpha Goat Polyclonal Antibody (Carboxyfluorescein)
Catalog # USBII8437-35F
Supplier:  US Biological
Anti-Interleukin 15 Receptor alpha Goat Polyclonal Antibody (Carboxyfluorescein)
Catalog # USBII8437-35F
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 15 Receptor Alpha
  • Antigen symbol:
    IL15RA
  • Clonality:
    Polyclonal
  • Conjugation:
    FAM (Carboxyfluorescein)
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Interleukin 15 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse