Anti-Interleukin 15 Receptor alpha Goat Polyclonal Antibody
Catalog # USBII8437-35E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 15 Receptor Alpha
- Antigen symbol:IL15RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Form:lyophilized powder
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Interleukin 15 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse