Order Entry
Export
ContactUsLinkComponent
Anti-Interleukin 10, Low Endotoxin Rat Monoclonal Antibody [clone: 9i107]
Anti-Interleukin 10, Low Endotoxin Rat Monoclonal Antibody [clone: 9i107]
Catalog # USBII8432-23D
Supplier:  US Biological
Anti-Interleukin 10, Low Endotoxin Rat Monoclonal Antibody [clone: 9i107]
Catalog # USBII8432-23D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 10
  • Antigen symbol:
    IL10
  • Clonality:
    Monoclonal
  • Clone:
    9i107
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:10-1:50; ; Western Blot: 1:100

Type: Primary
Antigen: Interleukin 10, Low Endotoxin
Clonality: monoclonal
Clone: 9i107
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human