Anti-Interferon lambda 2 Rabbit Polyclonal Antibody (Biotin)
50 µg
Catalog # USBII7662-35A-50
Supplier: US Biological
Anti-Interferon lambda 2 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBII7662-35A-50
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml
Type: Primary
Antigen: Interferon lambda 2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Form:lyophilized powder
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Concentration:~0,05 mg/ml
- Shipping temperature:Ice
- Antigen name:Interferon lambda 2
- Cat. No.:USBII7662-35A-50
- Reactivity:Human
- Conjugation:Biotin
- Antigen symbol:IFNL2
- Storage buffer:PBS
- ELISA:Yes



