Anti-Integrin, alpha4 Rabbit Polyclonal Antibody
Catalog # USBII7661-34A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin, alpha4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser988
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:10 Pieces
- Pk:10
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: Integrin, alpha4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human