Anti-IKBKG Mouse Monoclonal Antibody [clone: 6D477]
Catalog # USBII3000-36A2
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Inhibitor Of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase gamma
- Antigen symbol:IKBKG
- Clonality:Monoclonal
- Clone:6D477
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS containing 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: monoclonal
Clone: 6D477
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human