Anti-17-Hydroxypregnenolone Rabbit Polyclonal Antibody
USBIH9111-28Y
- Antibody type:Primary
- Antigen name:17-Hydroxypregnenolone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: 17-Hydroxypregnenolone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: