Anti-Hepatitis C Virus, E1 Antigen Mouse Monoclonal Antibody [clone: 10D100]
Catalog # USBIH1920-17D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis C Virus, E1 Antigen
- Clonality:Monoclonal
- Clone:10D100
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Hepatitis C Virus, E1 Antigen
Clonality: monoclonal
Clone: 10D100
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: