Anti-Granulocyte Macrophage Colony Stimulating Factor Rat Monoclonal Antibody [clone: 10B2332]
USBIG8951-07G
- Antibody type:Primary
- Antigen name:Granulocyte Macrophage Colony Stimulating Factor
- Antigen symbol:CSF2
- Clonality:Monoclonal
- Clone:10B2332
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:borate buffered saline
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Recommended dilution: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: Granulocyte Macrophage Colony Stimulating Factor
Clonality: monoclonal
Clone: 10B2332
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human