Order Entry
Export
ContactUsLinkComponent
 
Anti-G Protein Coupled Receptor 56 Rabbit polyclonal antibody
  USBIG8600-60B
Anti-G Protein Coupled Receptor 56 Rabbit polyclonal antibody
  USBIG8600-60B

 

  • Antibody type:
    Primary
  • Antigen name:
    G Protein Coupled Receptor 56
  • Antigen symbol:
    GPR56
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    buffer, 0,05% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Immunochemistry: 1:200

Type: Primary
Antigen: G Protein Coupled Receptor 56
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse