Anti-G Protein Coupled Receptor 51 Rabbit polyclonal antibody
USBIG8600-55C
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor 51
- Antigen symbol:GPR51
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser783
- Storage buffer:10mM HEPES, pH 7,5, 0,09% sodium azide, 0,01% BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:500
Type: Primary
Antigen: G Protein Coupled Receptor 51
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat