Anti-Glutamate Decarboxylase 65 Rabbit polyclonal antibody
Catalog # USBIG4000-01K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Glutamate decarboxylase 65
- Antigen symbol:GAD65
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:516 to 528
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Western Blot: 1:10-1:250
Type: Primary
Antigen: Glutamate Decarboxylase 65
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human