Anti-GSN Mouse Monoclonal Antibody [clone: 9j134]
Catalog # USBIG2024-52
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Gelsolin
- Antigen symbol:GSN
- Clonality:Monoclonal
- Clone:9j134
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:2000
Type: Primary
Antigen: GSN (gelsolin)
Clonality: monoclonal
Clone: 9j134
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human