Anti-FYN Mouse Monoclonal Antibody [clone: FYN-59]
Catalog # USBIF9500-03J
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:FYN oncogene related to SRC, FGR, YES
- Antigen symbol:FYN
- Clonality:Monoclonal
- Clone:FYN-59
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 15 ?g/ml
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: monoclonal
Clone: FYN-59
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human