Anti-Fucosyltransferase 11 Sheep Polyclonal Antibody
Catalog # USBIF9200-52
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fucosyltransferase 11
- Antigen symbol:FUT11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: Fucosyltransferase 11
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human