Anti-FOXP2 Rabbit Polyclonal Antibody
Catalog # USBIF9050-02
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box P2
- Antigen symbol:FOXP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,6mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1: 250000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: FOXP2 (forkhead box P2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human