Anti-Fetoprotein Mouse Monoclonal Antibody [clone: AFP-11]
Catalog # USBIF4100-02L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fetoprotein
- Clonality:Monoclonal
- Clone:AFP-11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by precipitation method
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Fetoprotein
Clonality: monoclonal
Clone: AFP-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human