Anti-PTK2 Mouse Monoclonal Antibody [clone: 0.T.43]
Catalog # USBIF0019-55-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:protein tyrosine kinase 2
- Antigen symbol:PTK2
- Clonality:Monoclonal
- Clone:0.T.43
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Rodent,Birds,Chicken,Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: monoclonal
Clone: 0.T.43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Avian, Chicken, Human, Rodent