Anti-Endoplasmic Reticulum Protein 57 Mouse monoclonal antibody [clone: 5H39]
Catalog # USBIE2291-75D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Endoplasmic Reticulum Protein 57
- Clonality:Monoclonal
- Clone:5H39
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:2000-1:5000
Type: Primary
Antigen: Endoplasmic Reticulum Protein 57
Clonality: monoclonal
Clone: 5H39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human