Order Entry
Export
ContactUsLinkComponent
Anti-Desmoglein 1,2 Mouse monoclonal antibody [clone: 5F189]
Anti-Desmoglein 1,2 Mouse monoclonal antibody [clone: 5F189]
Catalog # USBID3221-82W
Supplier:  US Biological
Anti-Desmoglein 1,2 Mouse monoclonal antibody [clone: 5F189]
Catalog # USBID3221-82W
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Desmoglein 1,2
  • Clonality:
    Monoclonal
  • Clone:
    5F189
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,4, 0,5% BSA, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,05 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Desmoglein 1,2
Clonality: monoclonal
Clone: 5F189
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat