Order Entry
Export
ContactUsLinkComponent
Anti-DDX6 Mouse monoclonal antibody [clone: 10B2681]
Anti-DDX6 Mouse monoclonal antibody [clone: 10B2681]
Catalog # USBID1660-45C
Supplier:  US Biological
undefined
Anti-DDX6 Mouse monoclonal antibody [clone: 10B2681]
Catalog # USBID1660-45C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DEAD (Asp-Glu-Ala-Asp) box helicase 6
  • Antigen symbol:
    DDX6
  • Clonality:
    Monoclonal
  • Clone:
    10B2681
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1 to 484
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Immunochemistry: 1. 5 ?g/ml; ELISA: 1 ?g/ml-3ng/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: DDX6 (DEAD (Asp-Glu-Ala-Asp) box helicase 6)
Clonality: monoclonal
Clone: 10B2681
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human