Anti-Cytokeratin 19 Fragment Mouse Monoclonal Antibody [clone: 11H24]
Catalog # USBIC9097-90
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 19 Fragment
- Clonality:Monoclonal
- Clone:11H24
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ion exchange chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Cytokeratin 19 Fragment
Clonality: Monoclonal
Clone: 11H24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human