Anti-Cytokeratin 18, Multiepitope Cocktail Mouse Monoclonal Antibody [clone: 5F223]
USBIC9097-23M
- Antibody type:Primary
- Antigen name:Cytokeratin 18, Multiepitope Cocktail
- Clonality:Monoclonal
- Clone:5F223
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Pig,Sheep,Mammals,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:5 ml
- Pk:5 mL
Type: Primary
Antigen: Cytokeratin 18, Multiepitope Cocktail
Clonality: Monoclonal
Clone: 5F223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Hamster, Human, Mammalian, Mouse, Pig, Rat, Sheep