Anti-Cysteine-rich, Angiogenic Inducer 61 Sheep Polyclonal Antibody
Catalog # USBIC9018-01H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cysteine-rich, Angiogenic Inducer 61
- Antigen symbol:CYR61
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Sheep,Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:22 to 381
- Storage buffer:0,2um filtered PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Cysteine-rich, Angiogenic Inducer 61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Sheep