Anti-Cysteine-rich, Angiogenic Inducer 61 Mouse Monoclonal Antibody [clone: 10C314]
Catalog # USBIC9018-01E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cysteine-rich, Angiogenic Inducer 61
- Antigen symbol:CYR61
- Clonality:Monoclonal
- Clone:10C314
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:250-1:1000
Type: Primary
Antigen: Cysteine-rich, Angiogenic Inducer 61
Clonality: Monoclonal
Clone: 10C314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human