Anti-Cysteine-rich, Angiogenic Inducer 61 Rabbit Polyclonal Antibody
Catalog # USBIC9018-01D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cysteine-rich, Angiogenic Inducer 61
- Antigen symbol:CYR61
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 1% BSA, 0,09% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:200
Type: Primary
Antigen: Cysteine-rich, Angiogenic Inducer 61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human