Anti-C-Peptide II Mouse Monoclonal Antibody [clone: 10B357]
Catalog # USBIC7905-26
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:C-Peptide II
- Clonality:Monoclonal
- Clone:10B357
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: C-Peptide II
Clonality: Monoclonal
Clone: 10B357
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat