Order Entry
Export
ContactUsLinkComponent
Anti-Cortistatin-29 Rabbit Polyclonal Antibody
Anti-Cortistatin-29 Rabbit Polyclonal Antibody
  USBIC7904-38K
Anti-Cortistatin-29 Rabbit Polyclonal Antibody
  USBIC7904-38K

 

  • Antibody type:
    Primary
  • Antigen name:
    Cortistatin 29
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    400 µg
  • Pk:
    400 µG

 

 

Type: Primary
Antigen: Cortistatin-29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human