Anti-Cortistatin-14 Rabbit Polyclonal Antibody
Catalog # USBIC7904-38A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cortistatin 14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Form:lyophilized powder
- Sequence:PCKNFFWKTFSSCK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:400 µg
- Pk:400 µG
Specifications
About this item
Type: Primary
Antigen: Cortistatin-14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat