Anti-Connexin 59 Rabbit Polyclonal Antibody (Biotin)
USBIC7859-59-BIOT
- Antibody type:Primary
- Antigen name:Connexin 59
- Antigen symbol:Cx59
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: Connexin 59
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human