Order Entry
Export
ContactUsLinkComponent
Anti-Connexin 59 Rabbit Polyclonal Antibody (Biotin)
Anti-Connexin 59 Rabbit Polyclonal Antibody (Biotin)
  USBIC7859-59A-BIOT
undefined
Anti-Connexin 59 Rabbit Polyclonal Antibody (Biotin)
  USBIC7859-59A-BIOT

 

  • Antibody type:
    Primary
  • Antigen name:
    Connexin 59
  • Antigen symbol:
    Cx59
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: Connexin 59
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse