Anti-Connexin 46 Rabbit Polyclonal Antibody
Catalog # USBIC7858-07C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Connexin 46
- Antigen symbol:Cx46
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,25 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-3?g/ml; Immunofluorescence: 3?g/ml; Immunochemistry: 3?g/ml
Type: Primary
Antigen: Connexin 46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse