Anti-Chorionic Gonadotropin, Human Mouse Monoclonal Antibody [clone: HCG-60]
Catalog # USBIC5069-02F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Chorionic gonadotropin
- Antigen symbol:hCG
- Clonality:Monoclonal
- Clone:HCG-60
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Chorionic Gonadotropin, Human
Clonality: Monoclonal
Clone: HCG-60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human