Anti-Cholecystokinin-33 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Catalog # USBIC5042-15A2
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cholecystokinin-33
- Clonality:Polyclonal
- Conjugation:FAM (Carboxyfluorescein)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Cholecystokinin-33
Clonality: Polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human