Anti-CEACAM 19 Mouse monoclonal antibody [clone: 9E84]
Catalog # USBIC2589-87
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CEACAM 19
- Clonality:Monoclonal
- Clone:9E84
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CEACAM 19
Clonality: Monoclonal
Clone: 9E+84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human