Anti-CD178 Rabbit Polyclonal Antibody
Catalog # USBIC2548-91H4
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD178
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:10mM PBS, pH 7,4, BSA, sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CD178
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human