Anti-MCAM Mouse Monoclonal Antibody [clone: SHM-57]
Catalog # USBIC2548-78W
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Melanoma Cell Adhesion Molecule
- Antigen symbol:MCAM
- Clonality:Monoclonal
- Clone:SHM-57
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:25 µg
- Pk:25 µG
Specifications
About this item
Type: Primary
Antigen: MCAM (melanoma cell adhesion molecule)
Clonality: Monoclonal
Clone: SHM-57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human