Anti-CD99 Mouse Monoclonal Antibody [clone: H036-1.1]
USBIC2444-02
- Antibody type:Primary
- Antigen name:CD99 molecule
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:H036-1.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CD99 (CD99 molecule)
Clonality: Monoclonal
Clone: H036-1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human