Anti-SELL Mouse Monoclonal Antibody [clone: DREG-56]
Catalog # USBIC2414-93S
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:DREG-56
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SELL (selectin L)
Clonality: Monoclonal
Clone: DREG-56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human