Anti-ICAM3 Mouse Monoclonal Antibody [clone: BL-LEUK- 50]
Catalog # USBIC2405-07D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:intercellular adhesion molecule 3
- Antigen symbol:ICAM3
- Clonality:Monoclonal
- Clone:BL-LEUK- 50
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Format:Supernatant
- Form:lyophilized powder
- Storage buffer:sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: ICAM3 (intercellular adhesion molecule 3)
Clonality: Monoclonal
Clone: BL-LEUK- 50
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: