Anti-ITGA5 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 5H10-27 (MFR5)]
Catalog # USBIC2405-01Y
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 5
- Antigen symbol:ITGA5
- Clonality:Monoclonal
- Clone:5H10-27 (MFR5)
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ITGA5 (integrin, alpha 5 (fibronectin receptor, alpha polypeptide))
Clonality: Monoclonal
Clone: 5H10-27 (MFR5)
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse