Anti-CD48 Mouse Monoclonal Antibody [clone: 5-4.8]
Catalog # USBIC2403-10A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD48 molecule
- Antigen symbol:CD48
- Clonality:Monoclonal
- Clone:5-4.8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:50mMsodium phosphate, pH 7,5, 100mM potassium chloride, 150mM sodium chloride
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CD48 (CD48 molecule)
Clonality: Monoclonal
Clone: 5-4.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human