Anti-CD45RA Mouse Monoclonal Antibody [clone: MRC OX-33]
Catalog # USBIC2400-30M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD45RA
- Clonality:Monoclonal
- Clone:MRC OX-33
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:125 µg
- Pk:125 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: Monoclonal
Clone: MRC OX-33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat