Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]
Catalog # USBIC2399-02K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:C11
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:50mMsodium phosphate, pH 7,5, 500mM potassium chloride, 150mM sodium chloride, 0,2% BSA, 0,04% sodium azide, 15% glycerol
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone: C11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human