Anti-CD44 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H170]
Catalog # USBIC2398-06D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD44 molecule (Indian blood group)
- Antigen symbol:CD44
- Clonality:Monoclonal
- Clone:7H170
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2, 5% glycerol, 0,2% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone: 7H170
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human