Anti-SPN Mouse Monoclonal Antibody [clone: MEM-59]
Catalog # USBIC2397-76V
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Sialophorin
- Antigen symbol:SPN
- Clonality:Monoclonal
- Clone:MEM-59
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SPN (sialophorin)
Clonality: Monoclonal
Clone: MEM-59
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human