Anti-CD40LG Rabbit Monoclonal Antibody [clone: 9E204]
USBIC2392-30E
- Antibody type:Primary
- Antigen name:CD40 ligand
- Antigen symbol:CD40LG
- Clonality:Monoclonal
- Clone:9E204
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,05% BSA, 0,01% sodium azide,50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:5000
Type: Primary
Antigen: CD40LG (CD40 ligand)
Clonality: Monoclonal
Clone: 9E+204
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat