Anti-CD9 Mouse Monoclonal Antibody [clone: CLB-47]
Catalog # USBIC2260-06L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD9 molecule
- Antigen symbol:CD9
- Clonality:Monoclonal
- Clone:CLB-47
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 10mg BSA, 0,001% merthiolate.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation and ion exchange chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: CD9 (CD9 molecule)
Clonality: Monoclonal
Clone: CLB-47
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human