Order Entry
Export
ContactUsLinkComponent
Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 53-6.7]
Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 53-6.7]
Catalog # USBIC2259-28G
Supplier:  US Biological
Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 53-6.7]
Catalog # USBIC2259-28G
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8a molecule
  • Antigen symbol:
    CD8A
  • Clonality:
    Monoclonal
  • Clone:
    53-6.7
  • Conjugation:
    PE (Phycoerythrin)/Cy5®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide and a stabilizing agent.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse